Learn More
NPS-2143 hydrochloride, MedChemExpress
Product Code. 16499956
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499956 50 mg 50mg
16473518 10 mg 10mg
16426247 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16499956 Supplier Medchem Express Supplier No. HY10171
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress NPS-2143 hydrochloride (SB-262470A hydrochloride), an orally active calcilytic agent, is a selective and potent calcium ion-sensing receptor (CaSR) antagonist. NPS-2143 hydrochloride (SB-262470A hydrochloride) blocks increases in cytoplasmic Ca2+ concentrations (IC50=43 nM) elicited by activating the Ca2+ receptor in HEK 293 cells expressing the human Ca2+ receptor.
NPS-2143 hydrochloride (SB-262470A hydrochloride), an orally active calcilytic agent, is a selective and potent calcium ion-sensing receptor (CaSR) antagonist. NPS-2143 hydrochloride (SB-262470A hydrochloride) blocks increases in cytoplasmic Ca2+ concentrations (IC50=43 nM) elicited by activating the Ca2+ receptor in HEK 293 cells expressing the human Ca2+ receptor.
Specifications
Chemical Name or Material NPS-2143 hydrochloride
CAS 324523-20-8
Color White
Purity Grade Notes Research
Molecular Formula C24H26Cl2N2O2
Quantity 50 mg
Synonym SB-262470A hydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (224.53 mM) ∣H2O : 1.85 mg/mL (4.15 mM; ultrasonic and warming and heat to 60°C)
SMILES N#CC1=C(OC[C@@H](CNC(C)(C)CC2=CC=C3C=CC=CC3=C2)O)C=CC=C1Cl.[H]Cl
Molecular Weight (g/mol) 445.38
Formula Weight 445.38
Percent Purity 99.94%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism