missing translation for 'onlineSavingsMsg'
Learn More

NPRB/NPR2 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18698100 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18698100 0.02 mL 0.02mL
18613550 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18698100 Supplier Novus Biologicals Supplier No. NBP2936650.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NPRB/NPR2 Polyclonal antibody specifically detects NPRB/NPR2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPRB/NPR2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias AMD1;AMDM;ANPb;ANPRB;Atrial natriuretic peptide receptor 2;ECDM;GCB;GC-B;GUC2B;GUCY2B;NPRB;NPRBi;SNSK
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 130-320 of human NPRB/NPR2 (NP_003986.2). SAKNDHYRTLVRTGPSAPKLGEFVVTLHGHFNWTARAALLYLDARTDDRPHYFTIEGVFEALQGSNLSVQHQVYAREPGGPEQATHFIRANGRIVYICGPLEMLHEILLQAQRENLTNGDYVFFYLDVFGESLRAGPTRATGRPWQDNRTREQAQALREAFQTVLVITYREPPNPEYQEFQNRLLIRARED
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, GPCR, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4882
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.