Learn More
NP118809, MedChemExpress
Product Code. 16446476
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
Product Code. 16446476
Brand: Medchem Express HY14462
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress NP118809 is a potent N-type calcium channel blocker, with an IC50 of 0.11 μM; also less potently inhibits L-type calcium channel with an IC50 of 12.2 μM.
NP118809 is a potent N-type calcium channel blocker, with an IC50 of 0.11 μM
Specifications
Chemical Name or Material NP118809
CAS 41332-24-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C32H32N2O
Quantity 100 mg
Synonym 39-1B4
Solubility Information DMSO : 50 mg/mL (108.55 mM; Need ultrasonic)
SMILES O=C(CC(C1=CC=CC=C1)C2=CC=CC=C2)N3CCN(C(C4=CC=CC=C4)C5=CC=CC=C5)CC3
Molecular Weight (g/mol) 460.61
Formula Weight 460.61
Percent Purity 98.79%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation