missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Noxa Antibody (69N15C9), Alexa Fluor™ 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-25266AF350
This item is not returnable.
View return policy
Description
Noxa Monoclonal specifically detects Noxa in Mouse samples. It is validated for Western Blot.
Specifications
| Noxa | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PMAIP1 | |
| This Noxa antibody was developed by immunizing mice with a fusion protein containing human Noxa. | |
| Protein G purified | |
| Apoptosis, Cancer, Cellular Markers, Membrane Trafficking and Chaperones, Necroptosis, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors | |
| 5366.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| 69N15C9 | |
| Western Blot | |
| adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa | |
| Rat | |
| 11.6 kDa | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction