missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The U-2 OS Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation, SDS-Page.
Specifications
Specifications
| For Use With (Application) | Immunoprecipitation, SDS-Page, Western Blot |
| Tissue | Bone/Cartilage |
| Quantity | 0.25 mg |
| Storage Buffer | HEPES (pH 7.9), KCl, Sucrose, Sodium deoxycholate, and NP-40 (without denaturing agents). |
| Purity | Protein |
| Test Specificity | U-2 OS (human osterogenic sarcoma) whole cell lysate (non-denatured). |
| Cellular Fraction | Whole |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?