missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ p19ARF/CDKN2A Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
Description
A peptide to CDKN2A.
Specifications
Specifications
| Gene ID (Entrez) | 12578 |
| Species | Human |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB 200-106. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | alternative reading frame, ARF, ARF-INK4a, CDK4I, CDKN2A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2 A, cyclin-dependent kinase inhibitor 2 A (p16, inhibits CDK4), cyclin-dependent kinase inhibitor 2 A, isoform 3, cyclin-dependent kinase inhibitor 2 A, isoforms 1/2, cyclin-dependent kinase inhibitor protein, INK4, INK4a, Ink4a/Arf, INK4a-ARF, mitochondrial smARF', MTS1, p14, p14ARF, P16, p16(INK4a), p16-INK4, p16INK4a, p16-INK4a, P19, p19 |
| Gene Symbol | Cdkn2a |
| Product Type | p19ARF |
| Quantity | 0.05 mg |
| Regulatory Status | RUO - research use only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction