missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The MCF-7 Cell Lysate is derived from human. It has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Cell Line | MCF-7 cell line lysate (breast cancer derived cells) |
| Quantity | 0.1 mg |
| Storage Buffer | 1X SDS sample buffer: 60mM Tris-HCl (pH 6.8) 10mM EDTA 2 % SDS 0.03% Bromophenol Blue 10% glycerol 0.3% Beta-mercaptoethanol |
| Host Species | Human |
| Content And Storage | -80°C in the dark. Avoid freeze-thaw cycles. |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?