Learn More
Novus Biologicals™ Recombinant Human TDP-43/TARDBP His Protein
Product Code. 18727953 Shop All Bio Techne Products
Click to view available options
Quantity:
0.02 mg
Unit Size:
0.02mg
This item is not returnable. View return policy
Product Code. 18727953
Brand: Novus Biologicals™ NBP222849
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-260 of Human TDP-43/TARDBP The Recombinant Human TDP-43/TARDBP Protein is derived from E. coli. The Recombinant Human TDP-43/TARDBP Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.02 mg