missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human DNAL1 His Protein

Product Code. 18167171 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18167171 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18167171 Supplier Novus Biologicals™ Supplier No. NBP223013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-190 of Human DNAL1 The Recombinant Human DNAL1 Protein is derived from E. coli. The Recombinant Human DNAL1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_113615
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA, 10% glycerol
Gene ID (Entrez) 83544
Molecular Weight (g/mol) 23.9kDa
Name DNAL1 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen DNAL1, 1-190aa Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMAKATTI KEALARWEEK TGQRPSEAKE IKLYAQIPPI EKMDASLSML ANCEKLSLST NCIEKIANLN GLKNLRILSL GRNNIKNLNG LEAVGDTLEE LWISYNFIEK LKGIHIMKKL KILYMSNNLV KDWAEFVKLA ELPCLEDLVF VGNPLEEKHS AENNWIEEAT KRVPKLKKLD GTPVIKGDEE EDN
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.