Learn More
Invitrogen™ Polypropylene Columns (Empty)
Click to view available options
Quantity:
50 Columns
Unit Size:
Pack of 50
Description
Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Format | Empty Column |
| Product Type | Polypropylene Column |
| Content And Storage | Store at room temperature. |
| Quantity | 50 Columns |