missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ NOV (Human) Recombinant Protein

Human NOV full-length ORF ( AAH15028, 1 a.a. - 357 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00004856-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138441

  • 508.00 € / 25µg
Estimated Shipment: 04-09-2024
to see stock.

Description

Description

  • Sequence: MQSVQSTSFCLRKQCLCLTFLLLHLLGQVAATQRCPPQCPGRCPATPPTCAPGVRAVLDGCSCCLVCARQRGESCSDLEPCDEGSGLYCDRSADPSKQTGICTAVEGDNCVFDGVIYRSGEKFQPSCKFQCTCRDGQIGCVPRCQLDVLLPEPNCPAPRKVEVPGECCEKWICGPDEEDSLGGLTLAAYRPEATLGVEVSDSSVNCIEQTTEWTACSKSCGMGFSTRVTNRNRQCEMLKQTRLCMVRPCEQEPEQPTDKKGKKCLRTKKSLKAIHLQFKNCTSLHTYKPRFCGVCSDGRCCTPHNTKTIQAEFQCSPGQIVKKPVMVIGTCTCHTNCPKNNEAFLQELELKTTRGKM
Specifications

Specifications

AAH15028
nephroblastoma overexpressed gene
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NOV
GST
4856
Wheat germ expression system
25 μg
CCN3, IGFBP9
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers