Learn More
Notopterol, TRC
Product Code. 30417069
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417069 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30417069 Supplier Toronto Research Chemicals Supplier No. TRCN88800010MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Notopterol
CAS 88206-46-6
Molecular Formula C21 H22 O5
Quantity 10 mg
Synonym 7H-Furo[3,2-g][1]benzopyran-7-one','4-[[(2E)-5-hydroxy-3,7-dimethyl-2,6-octadien-1-yl]oxy]-, 7H-Furo[3,2-g][1]benzopyran-7-one','4-[(5-hydroxy-3,7-dimethyl-2,6-octadienyl)oxy]-','(E)-(±)-, 7H-Furo[3,2-g][1]benzopyran-7-one','4-[[(2E)-5-hydroxy-3,7-dimethyl-2,6-octadienyl]oxy]- (9CI), 4-[[(2E)-5-Hydroxy-3,7-dimethyl-2,6-octadien-1-yl]oxy]-7H-furo[3,2-g][1]benzopyran-7-one, 7H-Furo[3,2-g][1]benzopyran-7-one','4-[(5-hydroxy-3,7-dimethyl-2,6-octadienyl)oxy]-','(E)-, Notopterol
InChI Formula InChI=1S/C21H22O5/c1-13(2)10-15(22)11-14(3)6-8-25-21-16-4-5-20(23)26-19(16)12-18-17(21)7-9-24-18/h4-7,9-10,12,15,22H,8,11H2,1-3H3/b14-6+
SMILES CC(=CC(O)C\C(=C\COc1c2C=CC(=O)Oc2cc3occc13)\C)C
IUPAC Name 4-[(2E)-5-hydroxy-3,7-dimethylocta-2,6-dienoxy]furo[3,2-g]chromen-7-one
Molecular Weight (g/mol) 354.4
Formula Weight 354.1467