missing translation for 'onlineSavingsMsg'
Learn More

Notoginsenoside Ft1, MedChemExpress

Product Code. p-200099555
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449188 10 mg 10mg
16441598 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16449188 Supplier Medchem Express Supplier No. HYN0910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Notoginsenoside Ft1 is a saponin isolated from Panax notoginseng; stimulator of angiogenesis.

Notoginsenoside Ft1 is a saponin isolated from Panax notoginseng

Specifications

Chemical Name or Material Notoginsenoside Ft1
CAS 155683-00-4
Color White
Purity Grade Notes Research
Molecular Formula C47H80O17
Quantity 10 mg
Solubility Information DMSO : 6.67 mg/mL (7.27 mM; Need ultrasonic)
SMILES C[C@@]([C@@]12C)(CC[C@@]3([H])C4(C)C)[C@@](C[C@@H](O)[C@]1([H])[C@]([C@](C)(O)CC/C=C(C)/C)([H])CC2)([H])[C@]3(CC[C@@H]4O[C@@](O[C@H](CO)[C@@H](O)[C@@H]5O)([H])[C@@H]5O[C@@](O[C@H](CO)[C@@H](O)[C@@H]6O)([H])[C@@H]6O[C@@](OC[C@@H](O)[C@@H]7O)([H])[C@@H]7O)C
Molecular Weight (g/mol) 917.13
Formula Weight 917.13
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.