Learn More
Invitrogen™ NOTCH4 Monoclonal Antibody (HMN4-14), PE, eBioscience™

Armenian Hamster Monoclonal Antibody
Brand: Invitrogen™ 12-5764-80
Description
Description: This HMN4-14 monoclonal antibody reacts with mouse Notch4, one of four members of the Notch family of receptors. Notch receptors are 300-kDa single-pass transmembrane proteins. While the extracellular domain contains numerous epidermal growth factor-like repeats for ligand binding, the intracellular domain is involved in cell signaling. Upon binding its membrane-bound ligand (either Delta or Jagged), the Notch receptor undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases the Notch intracellular domain (NICD), which subsequently translocates into the nucleus, forms a ternary complex with the DNA-binding transcription factor CSL (CBF-1, Su(H), Lag-1) and the coactivator Mastermind, and ultimately activates transcription. Notch 4 has been shown to be expressed in CD8+ splenic dendritic cells, endothelial cells (e.g. vascular smooth muscle cells), and macrophages. A truncated, constitutively active form of Notch 4 designated Int3 also exists in mice as a result of the mouse mammary tumor virus (MMTV) insertion into the Notch 4 gene. Composed of the transmembrane and intracellular domains of Notch 4, Int3 has been reported to be involved in mammary gland development and tumorigenesis in the mouse. Finally, studies have shown Notch 4 involvement in myeloid proliferation, hematopoiesis, and embryonic endothelial development. The HMN4-14 antibody is reported to also crossreact to rat and...
This gene encodes a member of the Notch family. Members of this Type 1 transmembrane protein family share structural characteristics including an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. Notch family members play a role in a variety of developmental processes by controlling cell fate decisions. This protein is cleaved in the trans-Golgi network, and presented on the cell surface as a heterodimer. This protein functions as a receptor for membrane bound ligands, and may play a role in vascular, renal and hepatic development. This gene may be associated with susceptibility to schizophrenia in a small portion of cases (entrez gene).
Specifications
| NOTCH4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P31695 | |
| NOTCH4 | |
| Affinity chromatography | |
| RUO | |
| 18132, 406162 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| HMN4-14 | |
| PE | |
| NOTCH4 | |
| DAQB-143M3.4; FLJ16302; hNotch4; Int3; Int-3; MGC74442; N4; neurogenic locus notch homolog protein 4; Notch 4; Notch 4 extracellular truncation; Notch 4 intracellular domain; Notch gene homolog 4; Notch homolog 4; notch receptor 4; NOTCH3; Notch4; putative Notch4 protein; Transforming protein Int-3 | |
| Armenian Hamster | |
| 25 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.