Learn More
NOTCH2/NOTCH2NL Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00 € - 349.00 €
Specifications
| Antigen | NOTCH2/NOTCH2NL |
|---|---|
| Clone | 2D5E1 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236754
|
Proteintech
68212-1-IG-20UL |
20 μL |
145.00 € 129.00 € / 20µL Save 16.00 € 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237970
|
Proteintech
68212-1-IG-150UL |
150 μL |
410.00 € 349.00 € / 150µL Save 61.00 € 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NOTCH2/NOTCH2NL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NOTCH2/NOTCH2NL. This antibody recognizes human antigen. The NOTCH2/NOTCH2NL antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| NOTCH2/NOTCH2NL | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 388677 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2D5E1 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| N2N | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |