missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notch-4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
463.00 €
Specifications
| Antigen | Notch-4 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Notch-4 Polyclonal specifically detects Notch-4 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Notch-4 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis | |
| PBS buffer, 2% sucrose | |
| 4855 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml, Immunohistochemistry | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| neurogenic locus notch homolog protein 4, notch 4, Notch homolog 4, NOTCH3 | |
| The immunogen is a synthetic peptide directed towards the middle region of human Notch-4. Peptide sequence LLLGLGAARELRDQAGLAPADVAHQRNHWDLLTLLEGAGPPEARHKATPG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title