missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOSTRIN Polyclonal Antibody
GREENER_CHOICE

Product Code. 15931495
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15931495 100 μg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15931495 Supplier Invitrogen™ Supplier No. PA572837

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NOSTRIN Polyclonal Antibody for Western Blot, IHC (P), ELISA

Multivalent adapter protein which may decrease NOS3 activity by inducing its translocation away from the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOSTRIN
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Nostrin
Gene Accession No. Q5I0D6, Q6WKZ7, Q8IVI9
Gene Alias BM247; BM247 homolog; Dab2-interacting protein 2; Daip2; Disabled homolog 2-interacting protein 2; endothelial nitric oxide synthase traffic inducer; ENOS traffic inducer; eNOS trafficking inducer; eNOS-trafficking inducer; mDaIP2; Nitric oxide synthase traffic inducer; nitric oxide synthase trafficker; nitric oxide synthase trafficking; nostrin; ortholog of mouse disabled 2 interacting proein 2; probable ortholog of human nitric oxide synthase trafficker (NOSTRIN)
Gene Symbols Nostrin
Host Species Rabbit
Immunogen NOSTRIN antibody for a 17 amino acid peptide near the carboxy terminus of human NOSTRIN. The immunogen is within amino acids 430 - 480 of NOSTRIN.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115677, 311111, 329416
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.