missing translation for 'onlineSavingsMsg'
Learn More

Nortadalafil, MedChemExpress

Product Code. 16464137
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16464137

Brand: Medchem Express HY90009

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nortadalafil is demethyl Tadalafil, which is a PDE5 inhibitor, currently marketed in pill form for treating erectile dysfunction (ED) under the name Cialis; and under the name Adcirca for the treatment of pulmonary arterial hypertension.

Nortadalafil is demethyl Tadalafil, which is a PDE5 inhibitor, currently marketed in pill form for treating erectile dysfunction (ED) under the name Cialis

Specifications

Chemical Name or Material Nortadalafil
CAS 171596-36-4
Color White
Purity Grade Notes Research
Molecular Formula C21H17N3O4
Quantity 50 mg
Synonym Demethyl Tadalafil
Solubility Information DMSO : 50 mg/mL (133.20 mM; Need ultrasonic)
SMILES O=C1NCC(N([C@@H]1C2)[C@H](C3=CC=C(OCO4)C4=C3)C5=C2C6=C(N5)C=CC=C6)=O
Molecular Weight (g/mol) 375.38
Formula Weight 375.38
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.