Learn More
Invitrogen™ Norovirus capsid G1 Monoclonal Antibody (4925)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547535
Description
Specificity: Specific for the GI capsid protein. Does not cross react with GII. Best Lateral Flow IA pair (capture/conjugate): Norovirus capsid G1 Monoclonal antibody (Product # MA5-47535)/Norovirus capsid G1 Polyclonal antibody (Product # MA5-18237). Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Specifications
| Norovirus capsid G1 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| ELISA, Lateral Flow, Western Blot | |
| 4925 | |
| Unconjugated | |
| Norwalk virus | |
| Proprietary sequence. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.