Learn More
Norgestimate metabolite Norelgestromin, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Norelgestromin is a metabolite of Norgestimate, which is a progestin or synthetic progestogen.
Specifications
Specifications
| Chemical Name or Material | Norgestimate metabolite Norelgestromin |
| CAS | 53016-31-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H29NO2 |
| Quantity | 5 mg |
| Synonym | 17-Deacetyl norgestimate 17-Deacylnorgestimate |
| Solubility Information | DMSO : 20 mg/mL (61.08 mM; Need ultrasonic) |
| SMILES | O/N=C1CC[C@@]2([H])C(CC[C@]3([H])[C@]2([H])CC[C@@]4(CC)[C@@]3([H])CC[C@]4(C#C)O)=C\1 |
| Molecular Weight (g/mol) | 327.46 |
| Show More |