missing translation for 'onlineSavingsMsg'
Learn More

NORE1 Antibody [Alexa Fluor« 488], Novus Biologicals Biologicals™

Product Code. 30498207 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30498207 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498207 Supplier Novus Biologicals Supplier No. NBP335254AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NORE1 Polyclonal antibody specifically detects NORE1 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NORE1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias Maxp1, MGC10823, MGC17344, New ras effector 1, NORE1A, NORE1B, NORE1RASSF3, Rap1-binding protein, RAPLnovel Ras effector 1, Ras association (RalGDS/AF-6) domain family 5, Ras association (RalGDS/AF-6) domain family member 5, ras association domain-containing protein 5, Ras effector-like protein, regulator for cell adhesion and polarization enriched in lymphoid tissue, Regulator for cell adhesion and polarization enriched in lymphoid tissues, tumor suppressor RASSF3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-265 of human NORE1 (NP_872606.1).,, Sequence:, MTVDSSMSSGYCSLDEELEDCFFTAKTTFFRNAQSKHLSKNVCKPVEETQRPPTLQEIKQKIDSYNTREKNCLGMKLSEDGTYTGFIKVHLKLRRPVTVPAGIRPQSIYDAIKEVNLAATTDKRTSFYLPLDAIKQLHISSTTTVSEVIQGLLKKFMVVDNPQKFALFKRIHKDGQVLFQKLSIADRPLYLRLLAGPDTEVLSFVLKENETGEVEWDAFSIPELQNFLTILEKEEQDKIQQVQKKYDKFRQKLEEALRESQGKPG
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 83593
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.