missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOR-1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17218453 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17218453 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17218453 Supplier Invitrogen™ Supplier No. PA5115669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NR4A3.

NOR-1, a NR4 nerve growth factor-like receptor, has been shown to affect neuronal development, T-cell apoptosis, and cancer development. NOR-1 binds to NBRE (NGFI-B response element) and regulates the pro-opiomelanocortin (POMC) gene. NOR-1 is induced rapidly but transiently by a variety of stimuli. Downregulation of NOR-1 and other NGFI-B family members is correlated with drug addiction-prone behavior of Lewis rats. Three human protein isoforms have been documented: NOR-1, NOR-1beta, and NOR-1 3' variant.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOR-1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene NR4A3
Gene Accession No. P51179, Q92570, Q9QZB6
Gene Alias AI573420; CHN; chondrosarcoma, extraskeletal myxoid, fused to EWS; CSMF; MINOR; mitogen-induced nuclear orphan receptor; Neuron-derived orphan receptor 1; neuron-derived orphan receptor 1/2; Nor1; NOR-1; NOR-2; Nr4a3; Nuclear hormone receptor NOR-1; nuclear hormone receptor NOR-1/NOR-2; Nuclear receptor subfamily 4 group A member 3; nuclear receptor subfamily 4, group A, member 3; Orphan nuclear receptor TEC; TEC; TEC receptor; Translocated in extraskeletal chondrosarcoma
Gene Symbols NR4A3
Host Species Rabbit
Immunogen A synthesized peptide derived from human NR4A3(Accession Q92570), corresponding to amino acid residues S390-L440.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18124, 58853, 8013
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.