missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nonphospho-4EBP1 (Thr46) Monoclonal Antibody (E.638.3)

Rabbit Monoclonal Antibody
Marque: Invitrogen™ MA515032
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
It is not recommended to aliquot this antibody.
This gene encodes one member of a family of translation repressor proteins. The protein directly interacts with eukaryotic translation initiation factor 4E, which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.
Spécification
| Nonphospho-4EBP1 (Thr46) | |
| Monoclonal | |
| 138 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| Q13541, Q60876, Q62622 | |
| EIF4EBP1 | |
| Synthetic peptide corresponding to residues surrounding Thr46 of human 4E-BP1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot | |
| E.638.3 | |
| Unconjugated | |
| EIF4EBP1 | |
| 4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116636, 13685, 1978 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido