Learn More
NONO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16887873 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16887873 20 μL 20µL
16877873 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16887873 Supplier Proteintech Supplier No. 110581AP20UL
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
This gene encodes an RNA-binding protein which plays various roles in the nucleus, including transcriptional regulation and RNA splicing. A rearrangement between this gene and the transcription factor E3 gene has been observed in papillary renal cell carcinoma. Alternatively spliced transcript variants have been described. Pseudogenes exist on Chromosomes 2 and 16.
Specifications
Antigen NONO
Applications Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NONO
Gene Accession No. Q15233, Q5FVM4, Q99K48
Gene Alias 55 kDa nuclear protein, NONO, NonO protein, NRB54, P54, p54(nrb), P54NRB
Gene Symbols Nono
Host Species Rabbit
Immunogen NONO Fusion Protein Ag1512
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317259, 4841, 53610
Target Species Mouse, Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG