Learn More
Nonafluoro-1-butanesulfonic Acid, TRC
Product Code. 17903605
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17903605 100 mg 100mg
17954175 1 g 1g
17992795 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17903605 Supplier Toronto Research Chemicals Supplier No. TRCN649325100MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 375-73-5
Molecular Formula C4 H F9 O3 S
Molecular Weight (g/mol) 300.1
Synonym 1-Butanesulfonic acid, nonafluoro- (6CI,7CI,8CI), 1,1,2,2,3,3,4,4,4-Nonafluoro-1-butanesulfonic acid, 1-Perfluorobutanesulfonic acid, Eftop FBSA, Nonafluoro-1-butanesulfonic acid, Nonafluorobutanesulfonic acid, Perfluorobutanesulfonic acid, PFBS
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonic acid
SMILES OS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C4HF9O3S/c5-1(6,3(9,10)11)2(7,8)4(12,13)17(14,15)16/h(H,14,15,16)
Formula Weight 299.9503
Chemical Name or Material Perfluorobutanesulfonic acid