missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Non-Vented Polystyrene Petri Dishes

Product Code. 10231301
Click to view available options
Type:
Heavy
Heavy Design
Standard
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 480
This item is not returnable. View return policy

Product Code. 10231301

Brand: Greiner Bio-One 632181

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for easy stacking

  • For long incubation periods
  • Manufactured from crystal polystyrene for optimal analysis by microscopy
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 80 mL
Color Clear
Material Polystyrene
Shape Round
Sterility Sterile
Type Standard
No. per Case 480
No. per Pack 20
Packaging Standard
Vent No
Height (Metric) 16 mm
Diameter (Metric) 94 mm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.