Learn More
NOMO2 Rabbit anti-Human, Polyclonal, Proteintech
Product Code. 16852914 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16852914 20 μL 20µL
16842914 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16852914 Supplier Proteintech Supplier No. 143281AP20UL
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
Specifications
Antigen NOMO2
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NOMO2
Gene Accession No. Q5JPE7
Gene Alias NODAL modulator 2, Nomo, NOMO2, PM5, pM5 protein 2
Gene Symbols NOMO2
Host Species Rabbit
Immunogen NOMO2 Fusion Protein Ag5595
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283820
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG