Learn More
Nomifensine Maleic Acid Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Nomifensine Hydrogen Maleate |
| CAS | 32795-47-4 |
| Molecular Formula | C16 H18 N2 . C4 H4 O4 |
| Quantity | 50 mg |
| Synonym | 1,2,3,4-Tetrahydro-2-methyl-4-phenyl-8-isoquinolinamine (2Z)-2-butenedioate (1:1), 1,2,3,4-Tetrahydro-2-methyl-4-phenyl-8-isoquinolinamine (Z)-2-butenedioate (1:1), (±)-Nomifensine hydrogen maleate, Alival, Alival (antidepressant), HOE 984, Hostalival, Merital, NSC 289114, Neurolene, Nomifensine hydrogen maleate, Nomifensine maleate, Psicronizer |
| InChI Formula | InChI=1S/C16H18N2.C4H4O4/c1-18-10-14(12-6-3-2-4-7-12)13-8-5-9-16(17)15(13)11-18;5-3(6)1-2-4(7)8/h2-9,14H,10-11,17H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | CN1CC(c2ccccc2)c3cccc(N)c3C1.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;2-methyl-4-phenyl-3,4-dihydro-1H-isoquinolin-8-amine |
| Molecular Weight (g/mol) | 354.4 |
| Formula Weight | 354.158 |