missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOD3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17208443 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17208443 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17208443 Supplier Invitrogen™ Supplier No. PA5115658

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NLRC3.

NOD3 is a member of the NOD (nucleotide-binding oligomerization domain) family, a group of proteins that are involved in innate immune defense. NOD3 also known as NLR family CARD containing 3 (NLRC3), is predominantly expressed in the immune system, particularly in T lymphocytes, and its expression is strongly down-regulated following stimulation of the T-cell receptor complex and CD28, suggesting that NOD3 plays a role in attenuating the activation of T cells. NOD3 inhibits NF-βB, AP-1 and NFAT transcriptional activation in Jurkat T cells downstream of CD3/CD28 stimulation or treatment with PMA/ionomycin and decreases IL-2 and CD25 mRNA induction in activated cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOD3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene NLRC3
Gene Accession No. Q5DU56, Q7RTR2
Gene Alias CARD15-like protein; caterpillar 16.2; Caterpiller 16.2; caterpiller protein 16.2; CLR16.2; D230007K08Rik; mFLJ00348; NACHT, LRR and CARD domains-containing protein 3; NLR family CARD domain containing 3; NLR family CARD domain-containing protein 3; NLR family, CARD domain containing 3; NLR family, CARD domain containing 3, pseudogene 1; Nlrc3; Nlrc3-ps1; NOD3; NOD-like receptor C3; Nucleotide-binding oligomerization domain protein 3; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 3; protein NLRC3; RGD1304902
Gene Symbols NLRC3
Host Species Rabbit
Immunogen A synthesized peptide derived from human NLRC3(Accession Q7RTR2), corresponding to amino acid residues L1026-M1065.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197358, 268857, 287070
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.