Learn More
Megro™ No. 18 Disposable Brown Needles
Description
- Precision tip according to ISO 9626
- Needle tube made of stainless steel
- Hub and protection cap made of polypropylene
- 0.45 x 25mm
- Provided 100/pk
Specifications
Specifications
| Certifications/Compliance | ISO 9626 |
| Material | Polypropylene (Hub), Stainless Steel (Barrel) |
| Color | Brown |
| Description | No. 18 |
| Length (Metric) | 0.45 x 25 mm |
| Disposable | Single-use |
| Quantity | 100/Pk. |