Learn More
R&D Systems™ NMS 873
Description
NMS 873 is a potent p97 ATPase (VCP) allosteric inhibitor (IC50 = 30 nM). Selective over all other AAA ATPases, HSP90 and kinases tested (IC50 >10 μM). Activates the unfolded protein response (UPR) and modulates autophagosome maturation. Exhibits antiproliferative activity in cancer cells in vitro (IC50 values are 400 and 700 nM for HCT116 and HeLa cells respectively). Reduces p97-sensitization to trypsin digestion.
Specifications
Specifications
| Chemical Name or Material | 3-[3-(Cyclopentylthio)-5-[[[2-methyl-4'-(methylsulfonyl)[1,1'-biphenyl]-4-yl]oxy]methyl]-4H-1,2,4-triazol-4-yl]pyridine |
| CAS | 1418013-75-8 |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Product Line | Tocris |
| Molecular Formula | C27H28N4O3S2 |
| Formula Weight | Observed MW: 520.67 |