Learn More
Invitrogen™ NMNAT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139759
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: TGRFIVIGGI VSPVHDSYGK QGLVSSRHRL IMCQLAVQNS DWIRVDPWEC YQDTWQTTCS VLEHHRDLMK RVTGCILSNV NTPSMTPVIG QPQNETPQPI YQNSNVATKP TAAKILGKVG ESLSRICCVR PPVERFTFVD ENANLGTVM Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 97%.
Catalyzes the formation of NAD(+) from nicotinamide mononucleotide (NMN) and ATP. Can also use the deamidated form; nicotinic acid mononucleotide (NaMN) as substrate but with a lower efficiency. Cannot use triazofurin monophosphate (TrMP) as substrate. Also catalyzes the reverse reaction, i.e. the pyrophosphorolytic cleavage of NAD(+). For the pyrophosphorolytic activity prefers NAD(+), NADH and NAAD as substrates and degrades nicotinic acid adenine dinucleotide phosphate (NHD) less effectively. Fails to cleave phosphorylated dinucleotides NADP(+), NADPH and NAADP(+).
Specifications
| NMNAT2 | |
| Polyclonal | |
| Unconjugated | |
| NMNAT2 | |
| AI843915; Blad; C1orf15; D030041I09Rik; KIAA0479; naMN adenylyltransferase 1; naMN adenylyltransferase 2; Nicotinamide mononucleotide adenylyltransferase 2; nicotinamide nucleotide adenylyltransferase 2; nicotinamide/nicotinic acid mononucleotide adenylyltransferase 2; nicotinate-nucleotide adenylyltransferase 1; nicotinate-nucleotide adenylyltransferase 2; NMN adenylyltransferase 2; NMN/NaMN adenylyltransferase 2; Nmnat2; PNAT1; PNAT2; Protein bloated bladder; pyridine nucleotide adenylyltransferase 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23057 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BZQ4 | |
| NMNAT2 | |
| Recombinant protein corresponding to Human NMNAT2. Recombinant protein control fragment (Product #RP-90847). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.