missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NME2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15915315
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15915315 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15915315 Supplier Invitrogen™ Supplier No. PA579744

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat heart tissue, rat brain tissue, mouse heart tissue, mouse brain tissue. IHC: Human Intestinal Cancer Tissue, Rat Cerebellum Tissue, mouse intestine tissue. ICC/IF: U20S cell, HELA Cell. Flow: HL-60 cell.

Nucleoside diphosphate kinase B is an enzyme that in humans is encoded by the NME2 gene. Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of 'A' (encoded by NME1) and 'B' (encoded by this gene) isoforms. Multiple alternatively spliced transcript variants encoding the same isoform have been found for this gene. Co-transcription of this gene and the neighboring upstream gene (NME1) generates naturally-occurring transcripts (NME1-NME2) which encode a fusion protein consisting of sequence sharing identity with each individual gene product.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NME2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene NME2
Gene Accession No. P19804, P22392, Q01768
Gene Alias 2.7.13.3; c-myc purine-binding transcription factor PUF; c-myc transcription factor; epididymis secretory sperm binding protein Li 155an; expressed in non-metastatic cells 2; expressed in non-metastatic cells 2 protein; HEL-S-155an; histidine protein kinase NDKB; NDK B; NDKB; NDP kinase alpha; NDP kinase B; NDPKB; NDPK-B; nm23-2; NM23B; nm23-H2; nm23-M2; NME/NM23 nucleoside diphosphate kinase 2; Nme2; non-metastatic cells 2, protein (NM23) expressed in; non-metastatic cells 2, protein (NM23B) expressed in; nucleoside diphosphate kinase B; nucleoside-diphosphate kinase 2; nucleotide diphosphate kinase B; P18; p18-12d; PUF
Gene Symbols NME2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human NME2 (137-152aa EELVDYKSCAHDWVYE).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18103, 4831, 83782
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.