missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NME1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15995305
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15995305 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15995305 Supplier Invitrogen™ Supplier No. PA579742

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human A549 whole cell, human MCF-7 whole cell, human Hela whole cell, human PC-3 whole cell, rat brain tissue.

NME1 was identified because of its reduced mRNA transcript levels in highly metastatic cells. Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of 'A' (encoded by this gene) and 'B' (encoded by NME2) isoforms. Mutations in the gene have been identified in aggressive neuroblastomas. Two transcript variants encoding different isoforms have been found for this gene. Co-transcription of this gene and the neighboring downstream gene (NME2) generates naturally-occurring transcripts (NME1-NME2), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NME1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene NME1
Gene Accession No. P15531, P15532, Q05982
Gene Alias AL024257; AWD; expressed in non-metastatic cells 1; expressed in non-metastatic cells 1 protein; expressed in non-metastatic cells 1 protein (NM23A) (nucleoside diphosphate kinase); expressed in non-metastatic cells 1, protein; expressed in non-metastatic cells 1, protein (NM23A) (nucleoside diphosphate kinase); GAAD; Granzyme A activated DNase (GAAD); Granzyme A-activated DNase; GZMA activated DNase; included; metastasis inhibition factor NM23; NB; NBR-A; NBR-B; NBS; NDK A; NDK A 1; NDK A 2; NDK NBR-A; NDK NBR-B; NDKA; NDP kinase A; NDP kinase A 1; NDP kinase A 2; NDP kinase beta; NDP kinase NBR-A; NDPKA; NDPK-A; NM23; NM23A; NM23-C1; NM23-H1; NM23H1B; nm23-M1; NME/NM23 nucleoside diphosphate kinase 1; Nme1; NME1-1; NME1-NME2 spliced read-through transcript; non-metastatic cells 1, protein (NM23A) expressed in; nucleoside diphosphate kinase A; nucleoside diphosphate kinase A 1; nucleoside diphosphate kinase A 2; nucleoside diphosphate kinase NBR-B; nucleoside-diphosphate kinase 1; nucleoside-diphosphate kinase NBR-A; nucleotide diphosphate kinase; nucloside diphosphate kinase; tumor metastatic process-associated protein
Gene Symbols NME1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human NM23A (137-152aa EELVDYTSCAQNWIYE).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18102, 191575, 4830
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.