missing translation for 'onlineSavingsMsg'
Learn More

NMDA, MedChemExpress

Product Code. p-200096717
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16427366

Brand: Medchem Express HY17551

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NMDA is a specific agonist for NMDA receptor mimicking the action of glutamate, the neurotransmitter which normally acts at that receptor.

NMDA is a specific agonist for NMDA receptor mimicking the action of glutamate, the neurotransmitter which normally acts at that receptor.

Specifications

Chemical Name or Material NMDA
Physical Form Powder
CAS 6384-92-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C5H9NO4
For Use With (Application) Neuroscience-Neurodegeneration
Synonym N-Methyl-D-aspartic acid
Solubility Information H2O : 33.33 mg/mL (226.53 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (67.97 mM; Need ultrasonic)
SMILES O=C(O)C[C@H](C(O)=O)NC
Molecular Weight (g/mol) 147.13
Formula Weight 147.13
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.