Learn More
Schmidlin™ NM Plus Rack Hydrogen Generator
Product Code. 17798919
Encompass_Preferred
Change view
Click to view available options
Flow Rate:
100 mL/min.
1000 mL/min.
250 mL/min.
500 mL/min.
Wattage:
90 W
125 W
200 W
400 W
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Wattage unitSize
17798919 100 mL/min. 90 W Each
17718929 250 mL/min. 125 W Each
17728929 500 mL/min. 200 W Each
17708929 1000 mL/min. 400 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17798919 Supplier Schmidlin™ Supplier No. DBRNM100EU
Only null left
Add to basket
Add to basket
This generator produces a continuous supply of high purity, dry hydrogen.
  • Increase efficiency and improve safety and performance
  • On-demand supply 24/7
  • Pressure: 11 barg (160 psig)
  • Unique permeation membrane drying system
  • Cold dual dynamic regeneration dryer
  • USB connectivity
  • Easy to install with complete 'plug and play' functionality
  • Ideal for all GC detector applications
  • Meets and exceeds requirements for the most demanding GC applications
  • Superior hydrogen production with reliable long life cell
  • Minimal maintenance; no desiccant cartridges to change
  • PC monitoring for maintenance, diagnostics and remote connection
Specifications
Depth (Metric) 500 cm
Flow Rate 100 mL/min.
Height (Metric) 13.3 cm
Max. Ambient Operating Temperature 35°C
Outlet Pressure 11 bar
Width (English) 19 in.
Width (Metric) 48.5 cm
Connections Outlet: 1/8 in.
Electrical Requirements 110/120 V at 60 Hz, 220/240 V at 50 Hz
Nitrogen Purity >99.99996%
Wattage 90 W