Learn More
Schmidlin™ NM Plus Rack Hydrogen Generator
Description
- Increase efficiency and improve safety and performance
- On-demand supply 24/7
- Pressure: 11 barg (160 psig)
- Unique permeation membrane drying system
- Cold dual dynamic regeneration dryer
- USB connectivity
- Easy to install with complete 'plug and play' functionality
- Ideal for all GC detector applications
- Meets and exceeds requirements for the most demanding GC applications
- Superior hydrogen production with reliable long life cell
- Minimal maintenance; no desiccant cartridges to change
- PC monitoring for maintenance, diagnostics and remote connection
Specifications
Specifications
| Depth (Metric) | 500 cm |
| Flow Rate | 500 mL/min. |
| Height (Metric) | 13.3 cm |
| Max. Ambient Operating Temperature | 35°C |
| Outlet Pressure | 11 bar |
| Width (English) | 19 in. |
| Width (Metric) | 48.5 cm |
| Connections | Outlet: 1/8 in. |
| Nitrogen Purity | >99.99996% |
| Wattage | 200 W |