missing translation for 'onlineSavingsMsg'
Learn More

NLRX1 Mouse anti-Human, Clone: OTI4H8, liquid, TrueMAB™

Product Code. 17522314
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17522314 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17522314 Supplier Origene Technologies Supplier No. TA809764

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NLR family member X1 (NLRX1) belongs to the NLRP family and contains four LRR (leucine-rich) repeats and one NATCH domain. NLRX1 localizes to the mitochondrial outer membrane and interacts through the mitochondrial antiviral signalling adaptor MAVS during viral infection. Expression of NLRX1 results in the potent inhibition of RLH- and MAVS-mediated interferon-beta promoter activity and in the disruption of virus-induced RLH-MAVS interactions. It is also reported that NLRX1 synergistically potentiated ROS production induced by tumour necrosis factor alpha, Shigella infection and double-stranded RNA, resulting in amplified NF-kappaB-dependent and JUN amino-terminal kinases-dependent signalling, thus contributing to the link between ROS generation at the mitochondria and innate immune responses.

Specifications

Antigen NLRX1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NLRX1
Gene Accession No. Q86UT6
Gene Alias CLR11.3, DLNB26, NOD26, NOD5, NOD9
Gene Symbols NLRX1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 866-975 of human NLRX1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79671
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.