Learn More
Invitrogen™ Nkx2.2 Monoclonal Antibody (3E4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517138
Description
MA5-17138 targets NKX2.2 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17138 immunogen is purified recombinant fragment of human NKX2.2 expressed in E. Coli. MA5-17138 detects NKX2.2 which has a predicted molecular weight of approximately 30.1kDa.
The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor.
Specifications
| Nkx2.2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O95096 | |
| NKX2-2 | |
| Purified recombinant fragment of human NKX2.2 expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 3E4 | |
| Unconjugated | |
| NKX2-2 | |
| Drosophila NK2 transcription factor related, locus 2; glycoprotein GP330, renal; homeobox protein NK-2 homolog B; Homeobox protein Nkx-2.2; homeobox protein Nkx2-2; NK2 homeobox 2; NK2 transcription factor related locus 2; NK2 transcription factor related, locus 2; NK2 transcription factor-like protein B; NKX2.2; Nkx-2.2; Nkx2\.2; Nkx2-2; NKX2B; tinman | |
| Mouse | |
| Protein G | |
| RUO | |
| 4821 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.