Learn More
Invitrogen™ NKRF Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5117006
Product Code. 17269503
  • 497.00 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Positive Samples: U-87MG, A-549, Mouse brain, Mouse spleen, Mouse kidney, Rat brain Immunogen sequence: YGTKKTSKHA AADEALKILQ KTQPTYPSVK SSQCHTGSSP RGSGKKKDIK DLVVYENSSN PVCTLNDTAQ FNRMTVEYVY ERMTGLRWKC KVILESEVIA EAVGVKKTVK YEAAGEAVKT LKKTQPTVIN NLKKGAVEDV ISRNEIQGRS AEEAYKQQIK EDNIGNQLLR KMGWTGGGLG KSGEGIREPI SVKEQHKREG LGLDVERVNK IAKRDIEQII RNYARSESHT DLTFSRELTN DERKQIHQIA QKYGLKSKSH GVGHDRYLVV GRKRRKEDLL DQLKQEGQVG HYELVMPQAN.
This gene encodes a transcription factor that interacts with specific negative regulatory elements (NREs) to mediate transcriptional repression of certain NK-kappa-B-responsive genes. The protein localizes predominantly to the nucleolus with a small fraction found in the nucleoplasm and cytoplasm.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NKRF
Polyclonal
Unconjugated
NKRF
9430034D17Rik; AW124770; C80287; ITBA4; ITBA4 protein; LOC100910054; NF-kappaB repressing factor; NF-kappa-B-repressing factor; NF-kappa-B-repressing factor-like; NFKB repressing factor; NFkB-repressing factor; NKRF; NRF; Protein ITBA4; Transcription factor NRF
Rabbit
Affinity chromatography
RUO
100910054, 55922, 77286
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.81 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15226, Q8BY02
NKRF
Recombinant fusion protein containing a sequence corresponding to amino acids 391-690 of human NKRF (NP_0600142).
100 μL
Primary
Human, Mouse, Rat