Learn More
Invitrogen™ NKRF Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117006
Description
Positive Samples: U-87MG, A-549, Mouse brain, Mouse spleen, Mouse kidney, Rat brain Immunogen sequence: YGTKKTSKHA AADEALKILQ KTQPTYPSVK SSQCHTGSSP RGSGKKKDIK DLVVYENSSN PVCTLNDTAQ FNRMTVEYVY ERMTGLRWKC KVILESEVIA EAVGVKKTVK YEAAGEAVKT LKKTQPTVIN NLKKGAVEDV ISRNEIQGRS AEEAYKQQIK EDNIGNQLLR KMGWTGGGLG KSGEGIREPI SVKEQHKREG LGLDVERVNK IAKRDIEQII RNYARSESHT DLTFSRELTN DERKQIHQIA QKYGLKSKSH GVGHDRYLVV GRKRRKEDLL DQLKQEGQVG HYELVMPQAN.
This gene encodes a transcription factor that interacts with specific negative regulatory elements (NREs) to mediate transcriptional repression of certain NK-kappa-B-responsive genes. The protein localizes predominantly to the nucleolus with a small fraction found in the nucleoplasm and cytoplasm.
Specifications
| NKRF | |
| Polyclonal | |
| Unconjugated | |
| NKRF | |
| 9430034D17Rik; AW124770; C80287; ITBA4; ITBA4 protein; LOC100910054; NF-kappaB repressing factor; NF-kappa-B-repressing factor; NF-kappa-B-repressing factor-like; NFKB repressing factor; NFkB-repressing factor; NKRF; NRF; Protein ITBA4; Transcription factor NRF | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100910054, 55922, 77286 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.81 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15226, Q8BY02 | |
| NKRF | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 391-690 of human NKRF (NP_0600142). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.