Learn More
Hofmann Klaus™ Nitrogen Schlenk Tube
Product Code. 10430991
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
Includes:
Standard Ground Cone
Standard Ground Socket
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Includes unitSize
10430991 10 mL Standard Ground Cone -
10547302 100 mL Standard Ground Cone -
10379700 10 mL Standard Ground Socket Each
10389700 25 mL Standard Ground Socket Each
10186580 50 mL Standard Ground Socket Each
10596912 25 mL Standard Ground Cone -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10430991 Supplier Hofmann Klaus™ Supplier No. 20511.29
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Description Nitrogen Tube with Side Stopcock, with Solid Glass Plug and Hose Connection
Capacity (Metric) 10 mL
Type Neck with Standard Ground Cone (14/23)
Includes Standard Ground Cone