missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic acid trisodium salt monohydrate, 98+%

Product Code. p-7024124
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11478730

Brand: Thermo Scientific Alfa Aesar A18348.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18662-53-8 | C6H8NNa3O7 | 275.099 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18662-53-8
Molecular Formula C6H8NNa3O7
Molecular Weight (g/mol) 275.099
MDL Number MFCD00150789
InChI Key HERBOKBJKVUALN-UHFFFAOYSA-K
Synonym trisodium nitrilotriacetate monohydrate, nitrilotriacetic acid trisodium salt monohydrate, nta sodium hydrate, nta trisodium salt dihydrate, unii-j0u92u6nu5, ccris 437, nitrilotriacetic acid trisodium salt, monohydrate, nitriloacetic acid trisodium salt monohydrate, glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate, nitrilotriacetic acid, trisodium salt monohydrate
PubChem CID 29194
IUPAC Name trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate
SMILES C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+]

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 3774022
Merck Index 14,6579
Formula Weight 275.12 (257.10 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Nitrilotriacetic acid trisodium salt monohydrate
compliance-icons compliance-icons
Product Identifier
  • Nitrilotriacetic acid trisodium salt monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Nitrilotriacetic acid trisodium salt monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.