missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ Nitrile-Coated Spandex Gloves with Aramid Fiber Lining

Product Code. 10263052
Change view
Click to view available options
:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10263052 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10263052 Supplier SHOWA™ Supplier No. 590010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Seamless knit gloves offer comfort and tactile sensitivity, with an oil-resistant coating for enhanced grip capabilities. Aramid fiber lining provides ANSI Cut Level 3 protection.

  • Stretchable, breathable spandex material
  • Oil-resistant textured coating provides enhanced grip, and tactile sensitivity
  • High puncture resistance

Specifications

ANSI Abrasion Rating ANSI 6
Construction Seamless Knit
Coating Material Nitrile
Color Yellow
Cut Resistance Rating ANSI A3
Exterior Finish Partially Textured
Hand Specific
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size Large
For Use With (Application) Glass Handling/Repairs, HVAC Systems, Steel Beams, and General Maintenance
Glove Cuff Style Knit
Puncture Rating ASTM F2878 LEVEL 1
Series 5900
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.