missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach Lange™ NitraVer X Nitrogen-Nitrate Reagent Set, HR
Click to view available options
Range:
0.2 to 30.0 mg/L NO3-N
Unit Size:
Each
Description
For determination of High Range Nitrate Nitrogen by the Chromotropic Acid Test 'N Tube Method. Hach Method 10020. Range: 0.2 - 30.0 mg/L NO3-N.50 tests per package.
- Test 'N Tube products provide safe, convenient testing
- Easy ordering
- Test 'N Tube products provide safe, convenient testing Capped 16-mm vials provide a self-contained package for mixing and measurement.
- Easy ordering All necessary reagents and vials are contained in the package.
- Description: NitraVer X Nitrate Reagent Set
- Method: 10020
- Method Name: Chromotropic Acid
- Number of tests: 50
- Parameter: Nitrate
- Platform: Test 'N Tube™
- Quantity: 50 /pk
- Range: 0.2 - 30.0 mg/L NO₃-N
- Special Remarks: Set contains: Cuvette test tubes, NitraVer X Reagent A (2605400), NitraVer X Reagent B Powder Pillows (2605546)
- Storage conditions: 10 - 25 °C
Specifications
Specifications
Specifications
| Range | 0.2 to 30.0 mg/L NO3-N |
| Type | Reagent Set |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction