missing translation for 'onlineSavingsMsg'
Learn More

Nitazoxanide, MedChemExpress

Product Code. 16464307
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464307 50 mg 50mg
16498418 10 mg 10mg
16468176 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16464307 Supplier Medchem Express Supplier No. HYB0217

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nitazoxanide (NTZ), an anthelmintic agent, exhibits a broad spectrum of activities against a wide variety of helminths, protozoa, and enteric bacteria infecting animals and humans.

Nitazoxanide inhibits Giardia lamblia trophozoite proliferation in axenic culture with an IC50 of 2.4 μM. Nitazoxanide can be used for the research of parasitic gastroenteritis. Nitazoxanide shows anti-Japanese encephalitis virus (JEV) activity in a mouse model.

Specifications

Chemical Name or Material Nitazoxanide
Physical Form Powder
CAS 55981-09-4
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C12H9N3O5S
For Use With (Application) COVID-19-anti-virus
Synonym NTZ NSC 697855
Solubility Information DMSO : ≥ 100 mg/mL (325.44 mM)
SMILES O=C(NC1=NC=C([N+]([O-])=O)S1)C2=CC=CC=C2OC(C)=O
Molecular Weight (g/mol) 307.28
Formula Weight 307.28
Percent Purity 98.35%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.