missing translation for 'onlineSavingsMsg'
Learn More

NIM1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15710421 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15710421 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15710421 Supplier Thermo Scientific Supplier No. PA552467

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

NIM1 Polyclonal Antibody

NIM1 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen NIM1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NIM1K
Gene Accession No. Q8IY84
Gene Alias E130304F04Rik; Nim1; NIM1 serine/threonine protein kinase; NIM1 serine/threonine-protein kinase; NIM1K; Serine/threonine-protein kinase NIM1
Gene Symbols NIM1K
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NIM1. Recombinant protein control fragment (Product #RP-89814).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 167359
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.