missing translation for 'onlineSavingsMsg'
Learn More

NIFK Polyclonal Antibody, Invitrogen™

Product Code. 15535601
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15535601 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15535601 Supplier Invitrogen Supplier No. PA538683

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NIFK is a nucleolar protein that interacts with Ki67, a protein strongly associated with, and a specific marker of, cell proliferation. The marsupial counterpart of human NIFK was recently identified and suggests that NIFK may be involved in the organization of higher order chromatin structure. NIFK was found to interact with the forkhead associated domain of Ki67 in mitotic cells and the interaction is thought to be phosphorylation-dependent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NIFK
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NIFK
Gene Accession No. Q9BYG3, Q91VE6
Gene Alias Ki67ip
Gene Symbols NIFK
Host Species Rabbit
Immunogen A synthetic peptide derived from the internal region of human NIFK
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67949, 84365
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.