Learn More
Nicotine Benzoate, TRC
Specifications
Specifications
| Chemical Name or Material | Nicotine Benzoate |
| CAS | 88660-53-1 |
| Molecular Formula | C10 H14 N2 . C7 H6 O2 |
| Quantity | 1 g |
| Synonym | Nicotine Benzoate, Benzoic acid','compd. with (S)-3-(1-methyl-2-pyrrolidinyl)pyridine (1:1), Pyridine','3-(1-methyl-2-pyrrolidinyl)-','(S)-','monobenzoate (9CI) |
| InChI Formula | InChI=1S/C10H14N2.C7H6O2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9;8-7(9)6-4-2-1-3-5-6/h2,4,6,8,10H,3,5,7H2,1H3;1-5H,(H,8,9)/t10-;/m0./s1 |
| SMILES | CN1CCC[C@H]1c2cccnc2.OC(=O)c3ccccc3 |
| IUPAC Name | benzoic acid;3-[(2S)-1-methylpyrrolidin-2-yl]pyridine |
| Molecular Weight (g/mol) | 284.353 |
| Formula Weight | 284.152 |