missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamide N-Methyltransferase/NNMT Antibody (OTI3D8), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18194607 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18194607 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18194607 Supplier Novus Biologicals Supplier No. NBP272984AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nicotinamide N-Methyltransferase/NNMT Monoclonal specifically detects Nicotinamide N-Methyltransferase/NNMT in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nicotinamide N-Methyltransferase/NNMT
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay
Classification Monoclonal
Clone OTI3D8
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 2.1.1.1, nicotinamide N-methyltransferase
Gene Symbols NNMT
Host Species Mouse
Immunogen Full length recombinant protein of human Nicotinamide N-Methyltransferase/NNMT (NP_006160) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4837
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.