Learn More
Nicotinamide-d4 (Major), TRC
Specifications
Specifications
| Chemical Name or Material | Nicotinamide-2,4,5,6-D4 |
| CAS | 347841-88-7 |
| Molecular Formula | C6 2H4 H2 N2 O |
| Quantity | 10 mg |
| Synonym | Nicotinamide D4 (2,4,5,6 D4), 3-Pyridine-2,4,5,6-d4-carboxamide, Nicotinamide-d4, Nicotinamide-2,4,5,6-D4, 3-(Aminocarbonyl)pyridine-2,4,5,6-d4 |
| InChI Formula | InChI=1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)/i1D,2D,3D,4D |
| SMILES | [2H]c1nc([2H])c(C(=O)N)c([2H])c1[2H] |
| IUPAC Name | 2,4,5,6-tetradeuteriopyridine-3-carboxamide |
| Molecular Weight (g/mol) | 126.15 |
| Formula Weight | 126.0731 |